Assay Details
Target Gene Details
Entrez Gene ID: | 234138 |
Gene Name: | TELO2 interacting protein 2 |
Gene Aliases: |
- |
Location: |
Chr.8:31150316-31164703 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tti2 | NM_001199988.1 | 6 | 1922 | NP_001186917.1 |
NM_144927.3 | 6 | 1922 | NP_659176.2 | |
NR_103719.1 | 6 | 1922 | ||
AK045392.1 | 6 | 1927 | BAC32341.1 | |
AK046031.1 | 6 | 1912 | BAC32576.1 | |
AK084242.1 | 6 | 1912 | BAC39146.1 | |
AK139425.1 | 6 | 1899 | BAE24007.1 | |
AK185341.1 | 1 | 39 | ||
AW120801.1 | 2 | 260 | ||
BC019943.1 | 6 | 1625 | AAH19943.1 |