Assay Details
Target Gene Details
Entrez Gene ID: | 12556 |
Gene Name: | cadherin 16 |
Gene Aliases: |
- |
Location: |
Chr.8:104601915-104624396 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdh16 | NM_001252627.1 | NP_001239556.1 | ||
NM_001252628.1 | NP_001239557.1 | |||
NM_007663.3 | NP_031689.1 | |||
AB009292.1 | ||||
AF016271.1 | AAC34254.1 | |||
AJ609635.1 | CAE82011.1 | |||
AK052636.1 | BAC35072.1 | |||
AK093905.1 | ||||
AK164546.1 | BAE37833.1 | |||
BC015251.1 | AAH15251.1 |
More Information
Set Membership: |
Intragenic Non-exonic |