Assay Details
Target Gene Details
Entrez Gene ID: | 11601 |
Gene Name: | angiopoietin 2 |
Gene Aliases: |
Agpt2, Ang-2, Ang2 |
Location: |
Chr.8:18690263-18741562 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Angpt2 | NM_007426.4 | NP_031452.2 | ||
AF004326.1 | AAB63189.1 | |||
AK019860.1 | BAB31887.1 | |||
AK048622.1 | BAC33396.1 | |||
AK143974.1 | ||||
AK156132.1 | BAE33599.1 | |||
BC027216.1 | AAH27216.1 | |||
BC138309.1 | ||||
BC138313.1 |
Target Gene Details
Entrez Gene ID: | 244329 |
Gene Name: | microcephaly, primary autosomal recessive 1 |
Gene Aliases: |
5430437K10Rik, BRIT1, D030046N04Rik, MCT |
Location: |
Chr.8:18595136-18803189 on Build GRCm38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mcph1 | NM_173189.2 | NP_775281.2 | ||
AK042026.1 | BAC31136.1 | |||
AY070216.1 | AAL50216.1 | |||
BC056924.1 | AAH56924.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |