Assay Details
Target Gene Details
Entrez Gene ID: | 69479 |
Gene Name: | RIKEN cDNA 1700029J07 gene |
Gene Aliases: |
- |
Location: |
Chr.8:45939368-45975454 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
1700029J07Rik | NM_001033148.3 | 5 | 2281 | NP_001028320.1 |
AK006501.1 | 6 | 875 | ||
AK156041.1 | 5 | 2283 | BAE33559.1 | |
BC107256.2 | 6 | 764 | AAI07257.1 |