Assay Details
Target Gene Details
Entrez Gene ID: | 66929 |
Gene Name: | anti-silencing function 1B histone chaperone |
Gene Aliases: |
1700003K02Rik, AA409591 |
Location: |
Chr.8:83955694-83970195 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Asf1b | NM_024184.2 | NP_077146.1 | ||
AK005646.1 | BAB24166.1 | |||
AK019594.1 | BAB31809.1 | |||
AK077005.1 | BAC36561.1 | |||
AK077718.1 | BAC36978.1 | |||
AK134906.1 | BAE22334.1 | |||
AK150818.1 | BAE29881.1 | |||
AK151951.1 | BAE30824.1 | |||
AK165322.1 | BAE38137.1 | |||
BC003428.1 | AAH03428.1 |
More Information
Set Membership: |
Intragenic Non-exonic |