Assay Details
Target Gene Details
Entrez Gene ID: | 11604 |
Gene Name: | agouti related neuropeptide |
Gene Aliases: |
Agrt, Art |
Location: |
Chr.8:105566695-105579845 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Agrp | NM_001271806.1 | NP_001258735.1 | ||
NM_007427.3 | NP_031453.1 | |||
BC079902.1 | AAH79902.1 | |||
CO044337.1 | ||||
U89484.1 | AAB68620.1 |
More Information
Set Membership: |
Intragenic Non-exonic |