Assay Details
Target Gene Details
Entrez Gene ID: | 69479 |
Gene Name: | RIKEN cDNA 1700029J07 gene |
Gene Aliases: |
- |
Location: |
Chr.8:45939368-45975454 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
1700029J07Rik |
Target Gene Details
Entrez Gene ID: | 212392 |
Gene Name: | coiled-coil domain containing 110 |
Gene Aliases: |
Gm172 |
Location: |
Chr.8:45934649-45945230 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc110 | NM_001033246.2 | NP_001028418.1 | ||
AK132742.1 | BAE21328.1 | |||
AK161248.1 | BAE36267.1 |
More Information
Set Membership: |
Intragenic Non-exonic |