Assay Details
Target Gene Details
Entrez Gene ID: | 74337 |
Gene Name: | paralemmin 3 |
Gene Aliases: |
4432412L15Rik, AL024300 |
Location: |
Chr.8:84021474-84030295 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Palm3 | NM_028877.1 | NP_083153.1 | ||
AK014493.1 | BAB29393.3 | |||
EF208034.1 |