Assay Details
Target Gene Details
Entrez Gene ID: | 75458 |
Gene Name: | chemokine-like factor |
Gene Aliases: |
1700001C14Rik, 1810018M11Rik, C32, CKLF1, CKLF3, CKLF4, CKLF5, Cklf2, Cklf6, HSPC224, UCK-1 |
Location: |
Chr.8:104250861-104264936 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cklf | NM_001037841.3 | 1 | 138 | NP_001032930.1 |
NM_001286383.1 | 1 | 138 | NP_001273312.1 | |
NM_029295.3 | 1 | 138 | NP_083571.1 | |
NR_104433.1 | 1 | 138 | ||
AF401530.1 | AAK94042.1 | |||
AF401531.2 | AAK94043.2 | |||
AK005577.1 | 1 | 98 | BAB24133.1 | |
AK032759.1 | 1 | 68 | ||
AK046397.1 | 1 | 90 | BAC32704.1 | |
AK050112.1 | 1 | 68 | BAC34072.1 | |
AK086063.1 | 1 | 69 | BAC39602.1 | |
AY046597.1 | 1 | 98 | AAL05945.1 | |
AY047360.1 | AAL10703.1 |
More Information
Set Membership: |
Intragenic Exonic |