Assay Details
Target Gene Details
Entrez Gene ID: | 70192 |
Gene Name: | CD209g antigen |
Gene Aliases: |
2310066I10Rik, AI481039, SIGNR7 |
Location: |
Chr.8:4134246-4137707 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd209g | NM_027343.3 | NP_081619.3 | ||
AK010071.1 | ||||
AM286437.1 | ||||
BC118542.1 | 1 | 2737 | AAI18543.1 |
More Information
Set Membership: |
Intragenic Non-exonic |