Assay Details
Target Gene Details
Entrez Gene ID: | 109006 |
Gene Name: | cytokine induced apoptosis inhibitor 1 |
Gene Aliases: |
2810413N20Rik, AA617265, AU021794, anamorsin |
Location: |
Chr.8:94819818-94838340 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ciapin1 | NM_134141.4 | 3 | 356 | NP_598902.1 |
AK076116.1 | 3 | 312 | BAC36196.1 | |
AK079615.1 | 3 | 318 | BAC37702.1 | |
AK088757.1 | 2 | 283 | BAC40550.1 | |
AK136821.1 | 3 | 350 | BAE23137.1 | |
AK146282.1 | 3 | 304 | BAE27040.1 | |
AK163705.1 | 3 | 335 | BAE37466.1 | |
AK168252.1 | 3 | 306 | BAE40202.1 | |
AK178553.1 | ||||
AK181039.1 | ||||
AK183681.1 | ||||
AK188252.1 | ||||
AK192608.1 | ||||
AK201030.1 | ||||
AK210999.1 | ||||
AK220040.1 | ||||
AY523555.1 | 2 | 277 | AAS09959.1 | |
BC016261.1 | 2 | 274 | AAH16261.1 | |
BC021864.1 | 3 | 296 | AAH21864.1 | |
BC021949.1 | 2 | 107 | AAH21949.1 | |
BY283649.1 |