Assay Details
Target Gene Details
Entrez Gene ID: | 20841 |
Gene Name: | zinc finger protein 143 |
Gene Aliases: |
AA959806, AU015869, D7Ertd805e, KRAB14, SBF, Staf, Zfp79, Zfp80-rs1, Znf143, pHZ-1 |
Location: |
Chr.7:110061670-110095396 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp143 | NM_009281.3 | 13 | 1562 | NP_033307.2 |
AK028056.1 | 13 | 1555 | BAC25726.1 | |
AK029078.1 | 13 | 1554 | BAC26281.1 | |
AK029926.1 | 13 | 1827 | BAC26682.1 | |
BC037658.1 | 12 | 1472 | AAH37658.1 |
More Information
Set Membership: |
Intragenic Exonic |