Assay Details
Target Gene Details
Entrez Gene ID: | 22228 |
Gene Name: | uncoupling protein 2 (mitochondrial, proton carrier) |
Gene Aliases: |
Slc25a8 |
Location: |
Chr.7:100493358-100499630 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ucp2 | NM_011671.5 | NP_035801.3 | ||
AK134116.1 | BAE22021.1 | |||
AK142687.1 | BAE25160.1 | |||
AK165245.1 | ||||
AK172307.1 | ||||
BC012697.1 | AAH12697.1 | |||
BY299894.1 | ||||
U69135.1 | AAB17666.1 | |||
U94593.1 | AAB53092.1 |
More Information
Set Membership: |
Intragenic Non-exonic |