Assay Details
Target Gene Details
Entrez Gene ID: | 17173 |
Gene Name: | achaete-scute complex homolog 2 (Drosophila) |
Gene Aliases: |
2410083I15Rik, Mash2, bHLHa45 |
Location: |
Chr.7:142966819-143009052 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ascl2 |
Target Gene Details
Entrez Gene ID: | 27027 |
Gene Name: | tetraspanin 32 |
Gene Aliases: |
AW208513, Art-1, BB235973, D7Wsu37e, Phemx, Tssc6 |
Location: |
Chr.7:143004902-143019486 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tspan32 | NM_001128080.2 | NP_001121552.1 | ||
AK041748.1 | BAC31052.1 | |||
AY039000.1 | AAK84430.1 |