Assay Details
Target Gene Details
Entrez Gene ID: | 56746 |
Gene Name: | testis expressed gene 101 |
Gene Aliases: |
1700008H15Rik, AI429076, TES101RP |
Location: |
Chr.7:24668012-24672050 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tex101 | NM_019981.2 | NP_064365.1 | ||
AB022914.1 | BAA90265.1 | |||
AK005769.1 | BAB24228.1 | |||
BC048475.1 | AAH48475.1 | |||
DQ222042.1 |