Assay Details
Target Gene Details
Entrez Gene ID: | 100503790 |
Gene Name: | predicted gene, 19897 |
Gene Aliases: |
- |
Location: |
Chr.7:28774899-28781514 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm19897 | NR_040339.1 | |||
AK040398.1 |
Target Gene Details
Entrez Gene ID: | 64383 |
Gene Name: | sirtuin 2 |
Gene Aliases: |
5730427M03Rik, SIR2L2, Sir2l |
Location: |
Chr.7:28766752-28788665 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sirt2 | NM_001122765.1 | NP_001116237.1 | ||
NM_001122766.1 | NP_001116238.1 | |||
NM_022432.4 | NP_071877.3 | |||
AB221570.1 | ||||
AF299337.1 | AAG39256.1 | |||
AK014042.1 | BAB29128.1 | |||
AK144431.1 | ||||
AK146412.1 | BAE27149.1 | |||
BB868649.1 | ||||
BC021439.1 | AAH21439.1 | |||
KF032392.1 |