Assay Details
Target Gene Details
Entrez Gene ID: | 232816 |
Gene Name: | zinc finger protein 628 |
Gene Aliases: |
Zec, Znf628 |
Location: |
Chr.7:4915176-4922003 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp628 | NM_170759.2 | NP_739565.2 | ||
AK048291.1 | ||||
AK155214.1 | BAE33125.1 | |||
BC056945.1 | AAH56945.1 | |||
BC062973.1 | AAH62973.1 | |||
BY202652.1 |
More Information
Set Membership: |
Intragenic Non-exonic |