Assay Details
Target Gene Details
Entrez Gene ID: | 16541 |
Gene Name: | napsin A aspartic peptidase |
Gene Aliases: |
KAP, Kdap, NAP1, SNAPA, pronapsin |
Location: |
Chr.7:44572445-44586846 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Napsa | NM_008437.1 | NP_032463.1 | ||
AB038144.1 | BAA90785.1 | |||
AK002520.1 | BAB22158.1 | |||
AK007861.1 | ||||
AK152663.1 | BAE31398.1 | |||
AK170013.1 | BAE41511.1 | |||
BC014813.1 | AAH14813.1 | |||
D88899.1 | BAA19004.1 |