Assay Details
Target Gene Details
Entrez Gene ID: | 72040 |
Gene Name: | cadherin-related family member 5 |
Gene Aliases: |
1810074H01Rik, AI481143, Mucdhl, Mupcdh |
Location: |
Chr.7:141269080-141276794 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdhr5 | NM_001114322.1 | NP_001107794.1 | ||
NM_028069.3 | NP_082345.1 | |||
AF462391.1 | AAL67856.1 | |||
AK007988.1 | BAB25392.1 | |||
AK027913.1 | BAC25662.1 | |||
BC043336.1 | ||||
BC054469.1 | AAH54469.1 |
More Information
Set Membership: |
Intragenic Non-exonic |