Assay Details
Target Gene Details
Entrez Gene ID: | 243967 |
Gene Name: | netrin 5 |
Gene Aliases: |
Gm484 |
Location: |
Chr.7:45684022-45694556 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ntn5 | NM_001033356.3 | NP_001028528.2 | ||
NM_001289692.1 | NP_001276621.1 | |||
NM_001289693.1 | NP_001276622.1 | |||
AK042348.1 | ||||
AK142898.1 | BAE25222.1 | |||
BC150901.1 |
Target Gene Details
Entrez Gene ID: | 56546 |
Gene Name: | secretory blood group 1 |
Gene Aliases: |
Fut10, Fut3 |
Location: |
Chr.7:45677686-45694402 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sec1 | NM_001271578.1 | NP_001258507.1 | ||
AK030246.1 | BAC26862.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |