Assay Details
Target Gene Details
Entrez Gene ID: | 67773 |
Gene Name: | K(lysine) acetyltransferase 8 |
Gene Aliases: |
2010203C02Rik, 5830450F21Rik, D7Ertd629e, MOF, Myst1 |
Location: |
Chr.7:127912517-127925763 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kat8 | NM_026370.1 | NP_080646.1 | ||
AK003264.1 | BAB22680.1 | |||
AK008418.1 | ||||
AK017161.1 | ||||
AK018002.1 | BAC25539.1 | |||
AK132000.1 | BAE20930.1 | |||
AK146710.1 | BAE27376.1 | |||
BC036284.1 | AAH36284.1 |
More Information
Set Membership: |
Intragenic Non-exonic |