Assay Details
Target Gene Details
Entrez Gene ID: | 56189 |
Gene Name: | proline dehydrogenase (oxidase) 2 |
Gene Aliases: |
2510028N04Rik, 2510038B11Rik, MmPOX, MmPOX1, POX1 |
Location: |
Chr.7:30493586-30513402 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prodh2 | NM_019546.5 | 2 | 293 | NP_062419.2 |
AF222851.1 | 2 | 280 | AAQ13907.1 | |
AK011020.1 | 2 | 284 | ||
AK011039.1 | 2 | 293 | ||
AK050141.1 | 2 | 294 | BAC34090.1 | |
BC018182.1 | 2 | 279 | AAH18182.1 | |
U80019.1 | 2 | 291 | AAF21466.1 |
More Information
Set Membership: |
Intragenic Exonic |