Assay Details
Target Gene Details
Entrez Gene ID: | 22342 |
Gene Name: | lin-7 homolog B (C. elegans) |
Gene Aliases: |
LIN-7B, MALS-2, Veli2 |
Location: |
Chr.7:45367891-45370564 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lin7b | NM_011698.1 | NP_035828.1 | ||
AF087694.1 | AAC78482.1 | |||
AF173082.1 | AAD48501.1 | |||
AK019299.1 | ||||
BC031780.1 | AAH31780.1 |
More Information
Set Membership: |
Intragenic Non-exonic |