Assay Details
Target Gene Details
Entrez Gene ID: | 84113 |
Gene Name: | prostate tumor over expressed gene 1 |
Gene Aliases: |
1110030G05Rik, AU041779, Gcap3 |
Location: |
Chr.7:44863067-44870411 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptov1 | NM_133949.1 | NP_598710.1 | ||
AK003974.1 | BAB23101.1 | |||
AK183935.1 | ||||
AK207550.1 | ||||
AK211543.1 | 1 | 218 | ||
AK211786.1 | 1 | 80 | ||
AK212557.1 | ||||
AK217872.1 | ||||
BC008543.1 | AAH08543.1 | |||
BC024632.1 | AAH24632.1 |
More Information
Set Membership: |
Intragenic Non-exonic |