Assay Details
Target Gene Details
Entrez Gene ID: | 213573 |
Gene Name: | calcium release activated channel regulator 2B |
Gene Aliases: |
6330520A15Rik, BC026645, Cracr2a, Efcab4, Efcab4a |
Location: |
Chr.7:141460874-141466613 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cracr2b | NM_001025103.2 | NP_001020274.2 | ||
BC026645.1 | AAH26645.1 | |||
BC034124.1 | AAH34124.1 | |||
BC048916.1 | AAH48916.1 | |||
BC098333.1 | AAH98333.1 |