Assay Details
Target Gene Details
Entrez Gene ID: | 13626 |
Gene Name: | embryonic ectoderm development |
Gene Aliases: |
ENSMUSG00000039373, l(7)5Rn, l7Rn5, lusk |
Location: |
Chr.7:89954654-89981419 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eed | NM_021876.3 | NP_068676.1 | ||
AK077664.1 | BAC36938.1 | |||
AK087924.1 | ||||
AK131976.1 | BAE20916.1 | |||
AK133890.1 | BAE21915.1 | |||
BC012966.1 | AAH12966.1 | |||
U78103.1 | AAB38319.1 | |||
U97675.1 | AAC53302.1 |
More Information
Set Membership: |
Intragenic Non-exonic |