Assay Details
Target Gene Details
Entrez Gene ID: | 232943 |
Gene Name: | kinesin light chain 3 |
Gene Aliases: |
BC017147, Klct |
Location: |
Chr.7:19394440-19404442 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klc3 | NM_001286038.1 | NP_001272967.1 | ||
NM_001286039.1 | NP_001272968.1 | |||
NM_146182.4 | NP_666294.1 | |||
AK158094.1 | BAE34354.1 | |||
BC017147.1 | AAH17147.1 |
More Information
Set Membership: |
Intragenic Non-exonic |