Assay Details
Target Gene Details
Entrez Gene ID: | 70973 |
Gene Name: | RIKEN cDNA 4931431B13 gene |
Gene Aliases: |
1700108I23Rik, AA536668 |
Location: |
Chr.7:128192050-128203328 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4931431B13Rik | NR_045183.1 | |||
AK007137.1 |
Target Gene Details
Entrez Gene ID: | 381924 |
Gene Name: | integrin, alpha D |
Gene Aliases: |
Cd11d |
Location: |
Chr.7:128173933-128205521 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 18 - Intron 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itgad | NM_001029872.3 | NP_001025043.3 | ||
AK156218.1 | BAE33629.1 |
More Information
Set Membership: |
Intragenic Non-exonic |