Assay Details
Target Gene Details
Entrez Gene ID: | 233875 |
Gene Name: | INO80 complex subunit E |
Gene Aliases: |
AI225782, AI854876, Ccdc85, Ccdc95 |
Location: |
Chr.7:126851584-126862371 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ino80e | NM_153580.1 | 1 | 161 | NP_705808.1 |
AK040862.1 | ||||
AK135531.1 | ||||
BC026475.1 | 1 | 161 | AAH26475.1 | |
BC055928.1 | AAH55928.1 |