Assay Details
Target Gene Details
Entrez Gene ID: | 100039953 |
Gene Name: | golgi-associated olfactory signaling regulator |
Gene Aliases: |
Gm2515, Gm581, Goofy |
Location: |
Chr.7:45176349-45179597 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gfy | NM_001195255.1 | 3 | 732 | NP_001182184.1 |
AB845286.1 | 1 | 459 | ||
BQ712971.1 | 3 | 615 | ||
BQ950069.1 | 3 | 687 |
Target Gene Details
Entrez Gene ID: | 114640 |
Gene Name: | parathyroid hormone 2 |
Gene Aliases: |
Tifp39, Tip39 |
Location: |
Chr.7:45176565-45181838 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pth2 |