Assay Details
Target Gene Details
Entrez Gene ID: | 100038912 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule pseudogene 1 |
Gene Aliases: |
Gm1978 |
Location: |
Chr.7:16649030-16657478 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ceacam-ps1 | NR_003247.2 |