Assay Details
Target Gene Details
Entrez Gene ID: | 69170 |
Gene Name: | RIKEN cDNA 1810026B05 gene |
Gene Aliases: |
- |
Location: |
Chr.7:73539798-73558395 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
1810026B05Rik | NR_037569.1 | 5 | 4024 | |
AK140938.1 | 5 | 4024 |
Target Gene Details
Entrez Gene ID: | 244059 |
Gene Name: | chromodomain helicase DNA binding protein 2 |
Gene Aliases: |
2810013C04Rik, 2810040A01Rik, 5630401D06Rik, AI851092, BC029703, CHD-2 |
Location: |
Chr.7:73426652-73541746 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Chd2 | NM_001081345.2 | NP_001074814.2 | ||
AK012735.1 | ||||
AK040559.1 | 1 | 831 | ||
AK088690.1 | ||||
AK132638.1 | BAE21274.1 | |||
AK133596.1 | BAE21742.1 | |||
AK136686.1 |