Assay Details
Target Gene Details
Entrez Gene ID: | 76932 |
Gene Name: | ADP-ribosylation factor interacting protein 2 |
Gene Aliases: |
2310002N04Rik, POR, Por1 |
Location: |
Chr.7:105634201-105640416 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arfip2 | NM_029802.4 | NP_084078.3 | ||
AK009100.1 | BAB26070.1 | |||
AK154772.1 | BAE32820.1 | |||
BC013794.1 | AAH13794.1 | |||
BC022942.1 | AAH22942.1 | |||
BC034520.1 | AAH34520.1 |
More Information
Set Membership: |
Intragenic Non-exonic |