Assay Details
Target Gene Details
Entrez Gene ID: | 13446 |
Gene Name: | double C2, alpha |
Gene Aliases: |
- |
Location: |
Chr.7:126847349-126852705 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Doc2a | NM_010069.1 | 3 | 972 | NP_034199.1 |
BC055768.1 | 3 | 972 | AAH55768.1 | |
D50000.1 | 2 | 395 | BAA23430.1 |