Assay Details
Target Gene Details
Entrez Gene ID: | 56787 |
Gene Name: | achaete-scute complex homolog 3 (Drosophila) |
Gene Aliases: |
Sgn1, bHLHa42 |
Location: |
Chr.7:109727465-109731732 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ascl3 | NM_020051.1 | NP_064435.1 | ||
AB046448.1 | BAB83911.1 | |||
AJ277605.1 | CAC37689.1 | |||
BC104346.1 | AAI04347.1 | |||
BC104347.1 | AAI04348.1 |
More Information
Set Membership: |
Intragenic Non-exonic |