Assay Details
Target Gene Details
Entrez Gene ID: | 333182 |
Gene Name: | cytochrome c oxidase subunit VIb polypeptide 2 |
Gene Aliases: |
1700067P11Rik, BC048670, COXVIB2 |
Location: |
Chr.7:4751792-4753094 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cox6b2 | NM_001289848.1 | 3 | 357 | NP_001276777.1 |
NM_001289849.1 | NP_001276778.1 | |||
NM_001289850.1 | NP_001276779.1 | |||
NM_183405.2 | 3 | 326 | NP_899664.1 | |
NM_183406.3 | 2 | 237 | NP_899665.1 | |
NR_110411.1 | 3 | 337 | ||
AK160152.1 | 3 | 256 | BAE35659.1 | |
AW123567.1 | ||||
AY152400.1 | 3 | 326 | AAN46753.1 | |
BC048670.1 | 2 | 237 | AAH48670.1 | |
BC107296.1 | 3 | 283 | AAI07297.1 | |
BC107297.1 | 3 | 283 | AAI07298.1 | |
BC125562.1 | 3 | 266 | ||
BC125564.1 | 3 | 269 | ||
BC145201.1 | 3 | 301 | ||
BG080371.2 | ||||
CT010366.1 | 2 | 170 | CAJ18573.1 |
More Information
Set Membership: |
Intragenic Exonic |