Assay Details
Target Gene Details
Entrez Gene ID: | 11425 |
Gene Name: | apolipoprotein C-IV |
Gene Aliases: |
Acl |
Location: |
Chr.7:19678084-19681460 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apoc4 | NM_007385.3 | 1 | 98 | NP_031411.1 |
BB872041.1 | 1 | 96 | ||
BC024657.1 | 1 | 54 | AAH24657.1 |
More Information
Set Membership: |
Intragenic Exonic |