Assay Details
Target Gene Details
Entrez Gene ID: | 11813 |
Gene Name: | apolipoprotein C-II |
Gene Aliases: |
- |
Location: |
Chr.7:19671579-19677941 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apoc2 | NM_001277944.1 | NP_001264873.1 | ||
AA059961.1 | ||||
AK146468.1 | BAE27195.1 | |||
BC024697.1 | AAH24697.1 | |||
BG085960.2 | ||||
CB949564.1 | ||||
Z15090.1 | CAA78804.1 |
More Information
Set Membership: |
Intragenic Non-exonic |