Assay Details
Target Gene Details
Entrez Gene ID: | 232791 |
Gene Name: | CCR4-NOT transcription complex, subunit 3 |
Gene Aliases: |
A930039N10Rik |
Location: |
Chr.7:3644522-3661109 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cnot3 | NM_146176.3 | NP_666288.1 | ||
AK141718.1 | 11 | 1713 | ||
AK173003.1 | BAD32281.1 | |||
BC030332.1 | AAH30332.1 | |||
BC053437.1 | AAH53437.1 | |||
BC141041.1 |
Target Gene Details
Entrez Gene ID: | 100628594 |
Gene Name: | microRNA 3572 |
Gene Aliases: |
mir-3572, mmu-mir-3572 |
Location: |
Chr.7:3655962-3656047 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mir3572 | NR_039587.1 | |||
JF446428.1 |
More Information
Set Membership: |
Intragenic Non-exonic |