Assay Details
Target Gene Details
Entrez Gene ID: | 21991 |
Gene Name: | triosephosphate isomerase 1 |
Gene Aliases: |
AI255506, Tpi, Tpi-1 |
Location: |
Chr.6:124810592-124814296 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tpi1 | NM_009415.2 | NP_033441.2 | ||
AK002270.1 | ||||
AK010808.1 | BAB27194.1 | |||
AK146013.1 | BAE26832.1 | |||
AK149768.1 | BAE29073.1 | |||
AK150735.1 | BAE29810.1 | |||
AK159741.1 | BAE35334.1 | |||
AK167437.1 | BAE39523.1 | |||
AK168060.1 | ||||
AK168446.1 | BAE40350.1 | |||
AK168756.1 | BAE40594.1 | |||
BC046761.1 | AAH46761.1 | |||
BU945752.1 | ||||
X53333.1 | CAA37420.1 |
More Information
Set Membership: |
Intragenic Non-exonic |