Assay Details
Target Gene Details
Entrez Gene ID: | 22377 |
Gene Name: | WW domain binding protein 1 |
Gene Aliases: |
AI255152 |
Location: |
Chr.6:83119044-83121663 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wbp1 | NM_001083922.1 | NP_001077391.1 | ||
NM_001083923.1 | NP_001077392.1 | |||
NM_016757.2 | NP_058037.2 | |||
AK076948.1 | BAC36533.1 | |||
AK152822.1 | ||||
AK170907.1 | 2 | 279 | ||
AK178117.1 | ||||
AK178563.1 | ||||
AK179139.1 | ||||
AK183702.1 | ||||
AK184383.1 | ||||
AK187014.1 | ||||
BC005666.1 | AAH05666.1 | |||
BC019889.1 | AAH19889.1 | |||
BE860581.1 | ||||
BY159385.1 | ||||
CX205200.1 | ||||
U40825.1 | AAB40892.1 |