Assay Details
Target Gene Details
Entrez Gene ID: | 22214 |
Gene Name: | ubiquitin-conjugating enzyme E2H |
Gene Aliases: |
1500009C23Rik, AI181839, AI326965, AI462483, AW227540, E2-20K, Gid3, N28148, Ubc8 |
Location: |
Chr.6:30211289-30304539 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ube2h | NM_001169576.1 | NP_001163047.1 | ||
NM_001169577.1 | NP_001163048.1 | |||
NM_009459.3 | NP_033485.1 | |||
AK131688.1 | BAE20763.1 | |||
AK161414.1 | BAE36382.1 | |||
BC008517.1 | AAH08517.1 | |||
U19854.1 | AAA91975.1 |
More Information
Set Membership: |
Intragenic Non-exonic |