Assay Details
Target Gene Details
Entrez Gene ID: | 27205 |
Gene Name: | podocalyxin-like |
Gene Aliases: |
AW121214, Ly102, PC, PCLP-1, Pclp1 |
Location: |
Chr.6:31519493-31563937 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Podxl | NM_013723.3 | 7 | 1517 | NP_038751.2 |
AB028048.1 | 7 | 1398 | BAA86912.1 | |
AF290209.1 | 7 | 1416 | AAG02458.1 | |
AK134723.1 | 3 | 253 | BAE22258.1 | |
BC052442.1 | 7 | 1491 | AAH52442.1 | |
BC054530.1 | 7 | 1491 | AAH54530.1 |
More Information
Set Membership: |
Intragenic Exonic |