Assay Details
Target Gene Details
Entrez Gene ID: | 14228 |
Gene Name: | FK506 binding protein 4 |
Gene Aliases: |
59kDa, AL022792, AW208983, FKBP-4, FKBP-52, FKBP52, FKPB52, p59 |
Location: |
Chr.6:128430107-128438631 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fkbp4 | NM_010219.3 | NP_034349.1 | ||
AK083912.1 | BAC39057.1 | |||
AK089972.1 | ||||
AK159211.1 | ||||
AK160202.1 | BAE35690.1 | |||
AK194969.1 | ||||
AK195473.1 | ||||
AK199911.1 | ||||
AK204158.1 | ||||
AK212190.1 | ||||
BC003447.1 | AAH03447.1 | |||
X17069.2 | CAC39452.1 | |||
X70887.1 | CAA50231.1 |
More Information
Set Membership: |
Intragenic Non-exonic |