Assay Details
Target Gene Details
Entrez Gene ID: | 74071 |
Gene Name: | intermediate filament tail domain containing 1 |
Gene Aliases: |
4933403M22Rik, AI482300, Lmna-rs1, Lmna1, Lmntd1 |
Location: |
Chr.6:145365142-145450766 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ifltd1 | NM_028742.2 | NP_083018.1 | ||
AK016641.1 | BAB30353.1 | |||
AK052206.1 | BAC34885.1 | |||
AY753326.1 | AAV31718.1 | |||
BC053420.1 | AAH53420.1 |