Assay Details
Target Gene Details
Entrez Gene ID: | 353172 |
Gene Name: | glycyl-tRNA synthetase |
Gene Aliases: |
GENA202, Gena201, Nmf249, Sgrp23 |
Location: |
Chr.6:55038001-55079504 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gars | NM_180678.3 | NP_851009.2 | ||
AK012758.2 | ||||
AK146238.1 | BAE27003.1 | |||
AK165857.1 | BAE38417.1 | |||
AK182981.1 | ||||
AK190167.1 | ||||
AK198707.1 | ||||
AK198907.1 | ||||
AK200032.1 | ||||
BC021747.1 | AAH21747.1 |
More Information
Set Membership: |
Intragenic Non-exonic |