Assay Details
Target Gene Details
Entrez Gene ID: | 74340 |
Gene Name: | S-adenosylhomocysteine hydrolase-like 2 |
Gene Aliases: |
4631427C17Rik, AI227036, Adohcyase3, mKIAA0828 |
Location: |
Chr.6:29768379-29912310 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 16 - Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ahcyl2 | NM_001171000.2 | NP_001164471.2 | ||
NM_001171001.1 | NP_001164472.1 | |||
NM_021414.6 | NP_067389.5 | |||
AK014539.1 | ||||
AK053527.1 | BAC35415.1 | |||
AK122382.1 | BAC65664.1 | |||
AK161892.1 | BAE36625.1 | |||
BC051504.1 | AAH51504.1 | |||
BC079660.1 | AAH79660.1 |
More Information
Set Membership: |
Intragenic Non-exonic |