Assay Details
Target Gene Details
Entrez Gene ID: | 66075 |
Gene Name: | coiled-coil-helix-coiled-coil-helix domain containing 3 |
Gene Aliases: |
0610041L09Rik, 1700039J09Rik, AW558177 |
Location: |
Chr.6:32792217-33060276 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Chchd3 | NM_025336.1 | 7 | 663 | NP_079612.1 |
AK002263.1 | 7 | 728 | BAB21974.1 | |
AK002889.1 | 7 | 734 | ||
AK003519.1 | 7 | 663 | BAB22832.1 | |
BC016528.1 | 6 | 485 | AAH16528.1 | |
BC021941.1 | 7 | 715 | AAH21941.1 |