Assay Details
Target Gene Details
Entrez Gene ID: | 66797 |
Gene Name: | contactin associated protein-like 2 |
Gene Aliases: |
5430425M22Rik, Caspr2, mKIAA0868 |
Location: |
Chr.6:45060061-47301388 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cntnap2 | NM_001004357.2 | 22 | 3870 | NP_001004357.2 |
NM_025771.3 | 2 | 317 | NP_080047.1 | |
AK013139.1 | 2 | 285 | BAB28674.1 | |
AK017341.1 | 2 | 312 | ||
AK129232.1 | 22 | 3843 | BAC98042.1 | |
BC029826.1 | 2 | 266 | AAH29826.1 | |
BC064467.1 | 17 | 2761 | AAH64467.1 | |
BC103799.1 | 2 | 224 | AAI03800.1 | |
BC132074.1 | 1 | 129 | ||
BC132076.1 | 1 | 129 | ||
BY297045.1 | 2 | 327 |