Assay Details
Target Gene Details
Entrez Gene ID: | 66797 |
Gene Name: | contactin associated protein-like 2 |
Gene Aliases: |
5430425M22Rik, Caspr2, mKIAA0868 |
Location: |
Chr.6:45060061-47301388 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cntnap2 | NM_001004357.2 | 20 | 3538 | NP_001004357.2 |
AK129232.1 | 20 | 3511 | BAC98042.1 | |
BC064467.1 | 15 | 2429 | AAH64467.1 |